Skip to Content
Merck
All Photos(1)

Key Documents

SRP2115

Sigma-Aldrich

RNA Polymerase II, p33 subunit, GST tagged human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

RPB3, RPB31, hRPB33, hsRPB3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,295.00

R$3,295.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
10 μG
R$3,295.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

R$3,295.00


Estimated to ship onApril 16, 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~59.1 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

150 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... POLR2C(5432)

Biochem/physiol Actions

In human, RPB3 is encoded by the POLR2C gene. This gene encodes the third largest subunit of RNA polymerase II. The product of this gene contains a cysteine rich region and exists as a heterodimer with another polymerase subunit, POLR2J. These two subunits form a core subassembly unit of the polymerase. DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Pol II is the central component of the basal RNA polymerase II transcription machinery. RPB3 is part of the core element with the central large cleft and the clamp element that moves to open and close the cleft. In the pol II complex, RPB3 interacts with RPB5 and the RPB3-RPB5 interaction is intensified in the presence of three subunits, RPB7, RPB8 and RPB11. Besides, RPB3 also interacts with RPB. RPB3 is involved in tissue-specific transcription and muscle differentiation via interaction with the myogenic factor Myogenin. RPB3 also interact with IGFBP3.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corbi, N., et al.
Faseb Journal, 10, 10963-10963 (2002)
J Acker et al.
The Journal of biological chemistry, 272(27), 16815-16821 (1997-07-04)
As an initial approach to characterizing the molecular structure of the human RNA polymerase II (hRPB), we systematically investigated the protein-protein contacts that the subunits of this enzyme may establish with each other. To this end, we applied a glutathione

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service