Skip to Content
Merck
All Photos(1)

Key Documents

SRP2096

Sigma-Aldrich

p300-Ch1 (302-531) human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

KAT3B, RP1-85F18.1, RSTS2, p300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~26 kDa

packaging

pkg of 10 μg

concentration

1000 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EP300(2033)

General description

The EP300 (E1A binding protein p300) gene is mapped to human chromosome 22q13.2. It codes for a histone acetyltransferase which is ubiquitously expressed.

Biochem/physiol Actions

Human p300 and CBP (CREB binding protein) are highly related transcriptional coactivators. Both proteins have been identified through protein interaction assays. In addition to interacting with variety of cellular factors and onco-proteins, loss of the wild type CBP alleles in isolated tumors suggests that CBP/p300 might serve as tumor suppressors. The ability of p300 to acetylate many transcription factors, including p53, E2F, TFIIE, and TFIIF etc. demonstrated a novel mechanism of targeted p300 regulation of gene expression.
Loss of EP300 (E1A binding protein p300) gene function causes Rubinstein-Taybi syndrome. p300 is essential for cell differentiation, growth and survival. p300 promotes liver stiffness, which is known to induce hepatic stellate cell differentiation to tumor-promoting myofibroblasts.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide sequencing expands the phenotypic spectrum of EP300 variants
Costain G, et al.
European Journal of Medical Genetics, 61(3), 125-129 (2018)
P300 acetyltransferase mediates stiffness-induced activation of hepatic stellate cells into tumor-promoting myofibroblasts
Dou C, et al.
Gastroenterology, 154(8), 2209-2221 (2018)
Dysregulation of Sirtuin 2 (SIRT2) and histone H3K18 acetylation pathways associates with adverse prostate cancer outcomes
Damodaran S, et al.
BMC Cancer, 17(1), 874-874 (2017)
Jeroen H Roelfsema et al.
American journal of human genetics, 76(4), 572-580 (2005-02-12)
CREB-binding protein and p300 function as transcriptional coactivators in the regulation of gene expression through various signal-transduction pathways. Both are potent histone acetyl transferases. A certain level of CREB-binding protein is essential for normal development, since inactivation of one allele
R Eckner et al.
Genes & development, 8(8), 869-884 (1994-04-15)
The growth-controlling functions of the adenovirus E1A oncoprotein depend on its ability ot interact with a set of cellular proteins. Among these are the retinoblastoma protein, p107, p130, and p300. We have isolated a cDNA encoding full-length human p300 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service