Skip to Content
Merck
All Photos(1)

Key Documents

SRP2066

Sigma-Aldrich

SRC1 (627-786), biotin, untagged human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

F-SRC-1, KAT13A, MGC129719, MGC129720, RIP160, SRC1, bHLHe42, bHLHe74

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
R$2,933.00

R$2,933.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 μG
R$2,933.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

R$2,933.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~19.8 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NCOA1(8648)

General description

Steroid receptor coactivator 1 (SRC1) is also known as nuclear receptor coactivator 1 (NCOA1). The gene encoding it is localized on human chromosome 2p23.3.[1]

Biochem/physiol Actions

Steroid receptor coactivator 1 (SRC1) is a transcriptional coactivator that mediates the activating functions of many of the nuclear hormone receptors. It is also known as NCoA1 and is a member of the SRC/p160 coactivator family. SRC1 has been shown to be over expressed in some cancers. SRC1 is a 160 kDa protein that contains several LXXLL motifs, which are involved in nuclear receptor interaction. The region 627-786 contains 3 LXXLL motifs that are involved in interaction with nuclear hormone receptorsand has been previously used in assays detecting ligand-dependent receptor-cofactor interactions. This protein does not contain a His tag and will be useful for assays where a native untagged protein is desired. Lane 1 corresponds to 6His-tagged and biotinylated SRC-RID; Lane 2 corresponds to biotinylated SRC-RID without 6His tag.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NCOA1 is a novel susceptibility gene for multiple myeloma in the Chinese population: A case-control study.
Peng M, et al.
PLoS ONE, 12(3), e0173298-e0173298 (2017)
Jianming Xu et al.
Molecular endocrinology (Baltimore, Md.), 17(9), 1681-1692 (2003-06-14)
The p160 steroid receptor coactivator (SRC) gene family contains three homologous members, which serve as transcriptional coactivators for nuclear receptors and certain other transcription factors. These coactivators interact with ligand-bound nuclear receptors to recruit histone acetyltransferases and methyltransferases to specific
S A Oñate et al.
Science (New York, N.Y.), 270(5240), 1354-1357 (1995-11-24)
A yeast two-hybrid system was used to identify a protein that interacts with and enhances the human progesterone receptor (hPR) transcriptional activity without altering the basal activity of the promoter. Because the protein stimulated transactivation of all the steroid receptors
D M Heery et al.
Nature, 387(6634), 733-736 (1997-06-12)
The binding of lipophilic hormones, retinoids and vitamins to members of the nuclear-receptor superfamily modifies the DNA-binding and transcriptional properties of these receptors, resulting in the activation or repression of target genes. Ligand binding induces conformational changes in nuclear receptors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service