Skip to Content
Merck
All Photos(1)

Key Documents

SRP0686

Sigma-Aldrich

TGFBR1 Peptide

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$845.00

R$845.00


Check Cart for Availability


Select a Size

Change View
1 MG
R$845.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

R$845.00


Check Cart for Availability

Product Name

TGFBR1 Peptide, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

lyophilized powder

mol wt

2116.45

shipped in

ambient

storage temp.

−20°C

Biochem/physiol Actions

The TGFBR1 peptide sequence (KKKVLTQMGSPSIRC-S(pS)VS) is derived from human SMAD3 (215-230) and is suitable for use as the substrate for TGFBR1 superfamily, including ACVRs (ALK1, ALK2, ALK4 and ALK7) and BMPRs (ALK3 and ALK6).

Sequence

KKKVLTQMGSPSIRC-S(pS)VS

Reconstitution

Reconstitute in 20mM Tris-HCl, pH 7.5 to a final concentration of 1mg/mL.

Storage and Stability

Store diluted product in aliquots at -20°C. Avoid freeze/thaw cycles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service