Skip to Content
Merck
All Photos(1)

Key Documents

SRP0648

Sigma-Aldrich

ARG1, His-Tag

recombinant, expressed in HEK 293 cells

Synonym(s):

Liver-Type Arginase 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$3,639.00

R$3,639.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
R$3,639.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

R$3,639.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

36 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ARG1(383)

General description

Human Arginase 1 (ARG1) protein, also known as LCN7 and LIECF3, GenBank Accession No. NM_000045, amino acids 1-322, with a C-terminal His-tag, expressed in an HEK293 cell expression system. MW = 36 kDa.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service