Skip to Content
Merck
All Photos(1)

Key Documents

SRP0466

Sigma-Aldrich

BRDT (22-138) His tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

BRD6, CT9, bromodomain, testis-specific

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,648.00

R$2,648.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μG
R$2,648.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$2,648.00


Estimated to ship onMay 28, 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

14.9 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRDT(676)

General description

Human Bromodomain testis-specific protein, also known as BRDT, BRD6, and CT9 (GenBank Accession No. NM_207189) amino-acids 22 – 138 with N-terminal HIS-tag, MW = 14.9 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binyamin D Berkovits et al.
Developmental biology, 360(2), 358-368 (2011-10-25)
Mice homozygous for a mutation (Brdt(∆BD1/∆BD1)) lacking the first bromodomain of Brdt, a testis-specific member of the BET family of double-bromodomain containing proteins, are sterile and exhibit profound defects in chromatin remodeling during spermiogenesis. We have now observed that a
Enyuan Shang et al.
Development (Cambridge, England), 134(19), 3507-3515 (2007-08-31)
Brdt is a testis-specific member of the distinctive BET sub-family of bromodomain motif-containing proteins, a motif that binds acetylated lysines and is implicated in chromatin remodeling. Its expression is restricted to the germ line, specifically to pachytene and diplotene spermatocytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service