Skip to Content
Merck
All Photos(1)

Key Documents

SRP0462

Sigma-Aldrich

BRD4 (342-460) His tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

CAP, HUNK1, HUNKI, MCAP, bromodomain containing 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,134.00

R$3,134.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
R$3,134.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$3,134.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

14.7 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRD4(23476)

General description

Human Bromodomain containing 4, also known as BRD4, HUNK1, or MCAP (GenBank Accession No. NM_058243) amino-acids 342-460 with N-terminal His-tag, MW = 14.7 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Voigt et al.
Genome biology, 12(11), 133-133 (2011-12-01)
The chromatin adapter BRD4 may be crucial for transmitting epigenetic information by acting as a histone acetylation-dependent gene bookmark and accelerating post-mitotic transcriptional reactivation.
R M Rodriguez et al.
Journal of molecular medicine (Berlin, Germany), 90(5), 587-595 (2011-11-29)
The bromodomain protein BRD4 is involved in cell proliferation and cell cycle progression, primarily through its role in acetylated chromatin-dependent regulation of transcription at targeted loci. Here, we show that BRD4 is frequently downregulated by aberrant promoter hypermethylation in human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service