Skip to Content
Merck
All Photos(2)

Documents

SRP0438

Sigma-Aldrich

TDP2 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥87% (SDS-PAGE)

Synonym(s):

EAP2, TTRAP, tyrosyl-DNA phosphodiesterase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥87% (SDS-PAGE)

form

aqueous solution

mol wt

67 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TDP2(51567)

General description

Human tyrosyl DNA phosphodiesterase 2, or TDP2 (GenBank Accession No. NM_016614) full length with N-terminal His-GST-tag, MW = 67 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Physical form

Formulated in 40 mM Tris-HCl, pH 8.0,110 mM NaCl, 2.2 mM KCl, 200 mM Imidazole, 20% Glycerol, and 3 mM DTT.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ka Cheong Lee et al.
International journal of molecular sciences, 19(7) (2018-07-18)
DNA topoisomerase II (TOP2) activity involves a normally transient double-strand break intermediate in which the enzyme is coupled to DNA via a 5'-phosphotyrosyl bond. However, etoposide and other topoisomerase drugs poison the enzyme by stabilising this enzyme-bridged break, resulting in
Christophe Marchand et al.
Molecular cancer therapeutics, 13(8), 2116-2126 (2014-07-16)
Drug screening against novel targets is warranted to generate biochemical probes and new therapeutic drug leads. TDP1 and TDP2 are two DNA repair enzymes that have yet to be successfully targeted. TDP1 repairs topoisomerase I-, alkylation-, and chain terminator-induced DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service