Skip to Content
Merck
All Photos(2)

Key Documents

SRP0430

Sigma-Aldrich

PDE4A4B active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥40% (SDS-PAGE)

Synonym(s):

DPDE2, PDE46, phosphodiesterase 4A, cAMP-specific

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,452.00

R$3,452.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
R$3,452.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$3,452.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥40% (SDS-PAGE)

form

aqueous solution

mol wt

124 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE4A(5141)

General description

Human PDE4A4B, also known as PDE4A isoform 1 (GenBank Accession No. NM_001111307) amino acids 2-end with N-terminal GST-tag, MW= 124 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole of 3?, 5?-cAMP to 5?-AMP per min at 37°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sullivan,M., et al.
Journal of Biochemistry, J. 333, 693-703 (1998)
Elaine Huston et al.
Journal of cell science, 119(Pt 18), 3799-3810 (2006-08-31)
The unique N-terminal regions of PDE4 cAMP-specific phosphodiesterases confer interaction with distinct signalling and scaffolding proteins. The PDE4A1 isoform is unique in being entirely membrane associated. Its N-terminal region is formed from two helices separated by a mobile hinge, where

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service