Skip to Content
Merck
All Photos(2)

Key Documents

SRP0390

Sigma-Aldrich

PRMT8 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

Histone-arginine N-methyltransferase 8, protein arginine N-methyltransferase 8

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$2,589.00

R$2,589.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
20 μG
R$2,589.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$2,589.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

40.8 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRMT8(56341)

General description

Human PRMT8 (protein arginine methyltransferase 8), (GenBank Accession No. NM_019854), amino acids 61-394 (end) with N-terminal His-FLAG-tag, MW=40.8 kDa, expressed in Sf9 cells using a Baculovirus expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuya Taneda et al.
Brain research, 1155, 1-9 (2007-05-22)
The regional distribution of PRMT8 transcript was examined in mouse brain using in situ hybridization (ISH) histochemistry. The PRMT8 cRNA probe was specifically hybridized with CNS and the signals were observed only in the neurons. The distribution of the neurons
Joyce Sayegh et al.
The Journal of biological chemistry, 282(50), 36444-36453 (2007-10-11)
Human protein arginine methyltransferase PRMT8 has been recently described as a type I enzyme in brain that is localized to the plasma membrane by N-terminal myristoylation. The amino acid sequence of human PRMT8 is almost 80% identical to human PRMT1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service