SRP0380
EZH2 /EED/SUZ12/RbAp48/AEBP2 human
recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)
About This Item
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥90% (SDS-PAGE)
form
aqueous solution
mol wt
48 kDa (RbBP5)
51 kDa (EED)
53 kDa (AEBP2)
86 kDa (EZH2)
87 kDa (SUZ12)
packaging
pkg of 20 μg
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... AEBP2(121536) , EED(8726) , EZH2(2146) , RBBP4(5928) , SUZ12(23512)
General description
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service