Skip to Content
Merck
All Photos(1)

Key Documents

SRP0273

Sigma-Aldrich

PDE8A1 Active human

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

Synonym(s):

PDE8, PDE8A, Phosphodiesterase 8A1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,134.00

R$3,134.00


Estimated to ship onMay 06, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
R$3,134.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$3,134.00


Estimated to ship onMay 06, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

specific activity

≥437 pmol/min-μg

mol wt

119 kDa

packaging

pkg of 10 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE8A(5151)

General description

PDE8A (phosphodiesterase 8A) is one of the two subfamilies of PDE8 protein family, the other being PDE8B. PDE8 shows wide range of tissue expression, and is especially abundant in testis.[1] PDE8A contains a Per, ARNT and Sim (PAS) signaling domain in its N-terminal. PDE8s contain receiver (REC) domain, N-glycosylation, N-myristoylation, amidation consensus sites, and potential kinase substrate sites for PKC (protein kinase), PKA and casein kinase.[2]
Human PDE8A1 (GenBank Accession No. NM_002605), full length with N-terminal GST tag, MW = 119kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

PDE8 (phosphodiesterase 8) are cAMP-specific enzymes, and they play a role in various processes such as in T-cell activation regulation, chemotaxis of activated lymphocytes, control of testosterone synthesis in Leydig cells, initiation of glucose-induced biphasic insulin response, steroidogenesis, and excitation–contraction coupling.[1][2] rs2304418 variant in this gene is linked with lower mRNA expression of PDE8A, which in turn is linked with reduced HIV-1 (human immunodeficiency virus) replication. The expression of this protein is significantly high in macrophages, and is controlled by miR-145-5p. PDE8A promotes HIV-1 replication in macrophages and this is controlled at the level of reverse transcription.[3][4]

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole of 3′, 5′-cAMP to 5′ AMP per min at 37°C

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphism in HIV-1 dependency factor PDE8A affects mRNA level and HIV-1 replication in primary macrophages.
Bol SM et al
Virology, 420(1), 32-42 (2011)
Cyclic AMP-specific phosphodiesterase, PDE8A1, is activated by protein kinase A-mediated phosphorylation.
Brown KM et al
Febs Letters, 586(11), 1631-1637 (2012)
Refolding and kinetic characterization of the phosphodiesterase-8A catalytic domain.
Yan Z et al
Protein Expression and Purification, 64(1), 82-88 (2009)
Thijs Booiman et al.
PloS one, 9(10), e109673-e109673 (2014-10-09)
HIV-1 infected macrophages play a key role in HIV-1 infection. Even during anti-retroviral treatment, macrophages keep producing virus due to suboptimal tissue penetration and reduced efficacy of antiretrovirals. It is therefore of major importance to understand which host factors are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service