Skip to Content
Merck
All Photos(1)

Key Documents

SRP0208

Sigma-Aldrich

PRL-1 (2-173) Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

PRL-1, PTP4A1, Protein tyrosine phosphatase type IVA, member 1, Protein-tyrosine phosphatase of regenerating liver 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,421.00

R$2,421.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
R$2,421.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

R$2,421.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

19.8 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTP4A1(7803)

General description

Human PRL 1 (GenBank Accession No. BC023975), (amino acids 2-173) with N-terminal HIS tag, MW = 19.8 kDa, expressed in an E. coli expression system.
Protein-tyrosine-phosphatase of regenerating liver 1 (PRL1), also known as protein tyrosine phosphatase type IVA, member 1 (PTP4A1), is encoded by the gene mapped to human chromosome 6q12. The encoded protein is a member of a small class of prenylated protein tyrosine phosphatases (PTPs). It is characterized by a PTP domain and a specific C-terminal prenylation motif. PRL1 is mainly localized to the cell membrane and the endoplasmic reticulum depending on its C-terminal prenylation.[1][2]

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Protein-tyrosine-phosphatase of regenerating liver 1 (PRL1) or protein tyrosine phosphatase type IVA, member 1 (PTP4A1) regulates cancer-related signaling pathways, which are downstream of epidermal growth factor receptor (EGFR) pathway and stimulates cell proliferation, invasion and migration. PRL1 acts as a potential therapeutic target and predictor for patient survival in prostate cancer (PCa) including those with castration-resistant prostate cancer (CRPC). PRL1 is essential for normal mitotic process. Elevated expressions of prenylated protein tyrosine phosphatases (PTPs) in mammalian cells conferred a transformed phenotype, which might be involved in the tumorigenesis.[2][3]

Unit Definition

One unit will hydrolyze 1 nmol p-nitrophenyl phosphate per minute at pH 7.4 and 30°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The gene encoding human nuclear protein tyrosine phosphatase, PRL-1. Cloning, chromosomal localization, and identification of an intron enhancer.
Peng Y, et al.
The Journal of Biological Chemistry, 273(27), 17286-17295 (1998)
Identification of PRL1 as a novel diagnostic and therapeutic target for castration-resistant prostate cancer by the Escherichia coli ampicillin secretion trap (CAST) method.
Shinmei S
Urologic Oncology, 32, 769-778 (2014)
Jing Wang et al.
The Journal of biological chemistry, 277(48), 46659-46668 (2002-09-18)
PRL-1 is one of three closely related protein-tyrosine phosphatases, which are characterized by C-terminal farnesylation. Recent reports suggest that they are involved in the regulation of cell proliferation and transformation. However, their biological function has not yet been determined. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service