Skip to Content
Merck
All Photos(1)

Key Documents

SRP0184

Sigma-Aldrich

Aha1 Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

AHA1, AHSA1, Activator of heat shock 90kDa protein ATPase homolog 1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
R$2,284.00

R$2,284.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
200 μG
R$2,284.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

R$2,284.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

38 kDa

packaging

pkg of 200 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AHSA1(10598)

General description

Human recombinant full-length AHA1 with C-terminal HIS tag (GenBank Accession No. NM_012111), expressed in E. coli. MW = 38 kDa,

Application

ATPase activity assay for Hsp90alpha.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 30% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service