Skip to Content
Merck
All Photos(1)

Key Documents

SML3997

Sigma-Aldrich

FOY-251 mesylate

≥98% (HPLC)

Synonym(s):

4-[[4-[(Aminoiminomethyl)amino]benzoyl]oxy]-benzeneacetic acid, methanesulfonate; 4-(4-Guanidinobenzoyloxy)phenylacetic acid, methanesulfonate, CC6 mesylate salt, Camostat active metabolite, FOY 251 mesylate salt, FOY 305 active metabolite, FOY-305 active metabolite, FOY251 mesylate salt, GBPA mesylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O4·CH4O3S
CAS Number:
Molecular Weight:
409.41
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(NC(N)=N)C=C1)OC2=CC=C(CC(O)=O)C=C2.O=S(O)(C)=O

InChI key

JXMOPIDYHUYOED-UHFFFAOYSA-N

Biochem/physiol Actions

Trypsin-like serine protease inhibitor Camostat (FOY-305) active metabolite with inhibitory potency against transmembrane serine protease 2 (TMPRSS2).



FOY-251 is a trypsin-like serine protease inhibitor and an active metabolite of the oral drug Camostat/FOY-305 (trypsin IC50 = 51/55 nM by FOY-305/FOY-251; plasma kallikrein/plasmin IC50 = 1.48/2.64 μM by FOY-305 and 2.32/12.5 μM by FOY-251). FOY-251 and FOY-305 are also potent inhibitor against transmembrane serine protease 2 (TMPRSS2 IC50 = 6.2/33.3 nM by by FOY-305/FOY-251) that primes SARS-CoV-2 spike protein for host cell entry.

Caution

Hygroscopic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New orally active serine protease inhibitors
Journal of medicinal chemistry, 38(14), 2521-2523 (1995)
Camostat mesylate inhibits SARS-CoV-2 activation by TMPRSS2-related proteases and its metabolite GBPA exerts antiviral activity
EBioMedicine, 65, 103255-103255 (2021)
Peptidomimetic inhibitors of TMPRSS2 block SARS-CoV-2 infection in cell culture
Communications biology, 5(1), 681-681 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service