Skip to Content
Merck
All Photos(1)

Key Documents

SML3932

Sigma-Aldrich

DC-Y13-27

≥98% (HPLC)

Synonym(s):

(E)-2-cyano-3-(5-(3-hydroxyphenyl)thiophen-2-yl)acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N2O2S
Molecular Weight:
270.31
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

room temp

SMILES string

NC(/C(C#N)=C/C1=CC=C(C2=CC=CC(O)=C2)S1)=O

InChI key

FCXTXGWIQGHISV-JXMROGBWSA-N

Biochem/physiol Actions

Cell penetrant and specific inhibitor of an m6A reader YTHDF2.



DC-Y13-27 is a cell penetrant and specific inhibitor of an m6A reader YTHDF2 that overcomes MDSC(myeloid-derived suppressor cell)-induced immunosuppression and enhances combined IR and/or anti-PD-L1 treatment.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liangliang Wang et al.
Cancer cell, 41(7), 1294-1308 (2023-05-27)
RNA N6-methyladenosine (m6A) modification is implicated in cancer progression. However, the impact of m6A on the antitumor effects of radiotherapy and the related mechanisms are unknown. Here we show that ionizing radiation (IR) induces immunosuppressive myeloid-derived suppressor cell (MDSC) expansion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service