Skip to Content
Merck
All Photos(1)

Documents

SML3595

Sigma-Aldrich

Riociguat

≥98% (HPLC)

Synonym(s):

BAY 63-2521, BAY 632521, Methyl [4,6-diamino-2-[1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-5-yl]methylcarbamate, N-[4,6-Diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]-N-methylcarbamic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19FN8O2
CAS Number:
Molecular Weight:
422.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

NC1=NC(C2=NN(CC3=CC=CC=C3F)C4=NC=CC=C42)=NC(N)=C1N(C(OC)=O)C

InChI

1S/C20H19FN8O2/c1-28(20(30)31-2)15-16(22)25-18(26-17(15)23)14-12-7-5-9-24-19(12)29(27-14)10-11-6-3-4-8-13(11)21/h3-9H,10H2,1-2H3,(H4,22,23,25,26)

InChI key

WXXSNCNJFUAIDG-UHFFFAOYSA-N

Biochem/physiol Actions

Riociguat is a soluble guanylate cyclase stimulator in clinical trials for pulmonary hypertension. It increases cGMP production and induces pulmonary vasodilation. It has not effect on phosphodiesterases PDE1-9 and PDE11 when used a concentrations less than 3µM.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 1 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riociguat for pulmonary hypertension--a glass half full
The New England Journal of Medicine, 369, 386-388 (2013)
Discovery of riociguat (BAY 63-2521): a potent, oral stimulator of soluble guanylate cyclase for the treatment of pulmonary hypertension
ChemMedChem, 4, 853-865 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service