Skip to Content
Merck
All Photos(1)

Documents

SML3468

Sigma-Aldrich

Pimpinellin

≥98% (HPLC)

Synonym(s):

5,6-Dimethoxy-2H-furo[2,3-h]-1-benzopyran-2-one, 6,7-Dimethoxyangelicin, Pimpinecilin, Pimpinelin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O5
CAS Number:
Molecular Weight:
246.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(C=CC1=C2OC)OC1=C3C(OC=C3)=C2OC

InChI

1S/C13H10O5/c1-15-11-7-3-4-9(14)18-10(7)8-5-6-17-12(8)13(11)16-2/h3-6H,1-2H3

InChI key

BQPRWZCEKZLBHL-UHFFFAOYSA-N

Biochem/physiol Actions

Pimpinellin is a coumarin isolated from roots of Angelica dahurica and Zosima absinthifolia that exhibits anti-tumor, anti-allergic, antioxidant and anticholinesterase activities. It appears to reduce the release of histamine, secretion of TBF‑α, interleukin (IL)‑1β and IL‑4. Pimpinellin exhibits potent cytotoxic activities against MGC-803, PC3, and A375 cancer cell lines.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service