Skip to Content
Merck
All Photos(2)

Key Documents

SML3399

Sigma-Aldrich

CHZ868

≥98% (HPLC)

Synonym(s):

CHZ 868, CHZ-868, N-(4-((2-((2,4-Difluorophenyl)amino)-1,4-dimethyl-1H-benzo[d]imidazol-5-yl)oxy)pyridin-2-yl)acetamide, N-[4-[[2-[(2,4-Difluorophenyl)amino]-1,4-dimethyl-1H-benzimidazol-5-yl]oxy]-2-pyridinyl]acetamide, NVP-CHZ 868, NVP-CHZ-868, NVP-CHZ868

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19F2N5O2
CAS Number:
Molecular Weight:
423.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to pink

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Orally active, type II tyrosine kinase I inhibitor against VEGFR1/2 (FLT1/KDR), PDGFR1/2, KIT, and ROS1 oncogenic fusions/mutations-drivent growth.

CHZ868 is a type II tyrosine kinase inhibitor that exhibits potent antiproliferation activity against VEGFR1/2 (FLT1/KDR), PDGFR1/2, KIT, and ROS1 TEL fusions-, as well as JAK2 V617F, but not RAF V600E, mutation-driven oncogenic growth (IC50 = 2/11, 4/23, 16, 40, 51 against respective BaF3 transformants) via stablizing target kinases in the inactive “DFG-out” conformation. CHZ868 overcomes Type I JAK inhibitor resistance of JAK2/MPN mutant MPN and JAK2 I682F/CRLF2-rearranged B-ALL cells both in cultures and in mice in vivo (30-40 mg/kg/day p.o.). Comparing to BBT594, CHZ868 is ineffective against BCR-ABL, TEL-FLT3, TEL-RET.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

miR-204 inhibits angiogenesis and promotes sensitivity to cetuximab in head and neck squamous cell carcinoma cells by blocking JAK2-STAT3 signaling
Biomedicine and Pharmacotherapy, 99, 278-285 (2018)
How Does the L884P Mutation Confer Resistance to Type-II Inhibitors of JAK2 Kinase: A Comprehensive Molecular Modeling Study
Scientific Reports, 7(1), 9088-9088 (2017)
PDGFRB mutation and tyrosine kinase inhibitor resistance in Ph-like acute lymphoblastic leukemia
Blood, 131(20), 2256-2261 (2018)
CHZ868, a Type II JAK2 Inhibitor, Reverses Type I JAK Inhibitor Persistence and Demonstrates Efficacy in Myeloproliferative Neoplasms
Cancer Cell, 28(1), 15-28 (2015)
Ah Ram Kim et al.
Cell, 168(6), 1053-1064 (2017-03-12)
Cytokines are classically thought to stimulate downstream signaling pathways through monotonic activation of receptors. We describe a severe anemia resulting from a homozygous mutation (R150Q) in the cytokine erythropoietin (EPO). Surprisingly, the EPO R150Q mutant shows only a mild reduction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service