Skip to Content
Merck
All Photos(1)

Key Documents

SML3364

Sigma-Aldrich

Emodepside

≥95% (HPLC)

Synonym(s):

Bay-44-4400, Cyclo[(αR)-α-hydroxy-4-(4-morpholinyl)benzenepropanoyl-N-methyl-L-leucyl-(2R)-2-hydroxypropanoyl-N-methyl-L-leucyl-(αR)-α-hydroxy-4-(4-morpholinyl)benzenepropanoyl-N-methyl-L-leucyl-(2R)-2-hydroxypropanoyl-N-methyl-L-leucyl], PF 1022-221

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H90N6O14
CAS Number:
Molecular Weight:
1119.39
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

N5(CCOCC5)c1ccc(cc1)C[C@H]2OC(=O)[C@@H](N(C(=O)[C@H](OC(=O)[C@@H](N(C(=O)[C@H](OC(=O)[C@@H](N(C(=O)[C@H](OC(=O)[C@@H](N(C2=O)C)CC(C)C)C)C)CC(C)C)Cc3ccc(cc3)N4CCOCC4)C)CC(C)C)C)C)CC(C)C

InChI

1S/C60H90N6O14/c1-37(2)31-47-57(71)77-41(9)53(67)61(11)50(34-40(7)8)60(74)80-52(36-44-17-21-46(22-18-44)66-25-29-76-30-26-66)56(70)64(14)48(32-38(3)4)58(72)78-42(10)54(68)62(12)49(33-39(5)6)59(73)79-51(55(69)63(47)13)35-43-15-19-45(20-16-43)65-23-27-75-28-24-65/h15-22,37-42,47-52H,23-36H2,1-14H3/t41-,42-,47+,48+,49+,50+,51-,52-/m1/s1

InChI key

ZMQMTKVVAMWKNY-YSXLEBCMSA-N

Biochem/physiol Actions

Cyclooctadepsipeptide anthelmintic
Emodepside is a cyclooctadepsipeptide anthelmintic a novel mode of action involving the calcium-gated potassium channel SLO-1 in C. elegans, sometimes mediated by a latrophilin (LAT) receptor.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT RE 2

Target Organs

Pancreas,Adrenal gland

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service