SML3356
Gefarnate
≥98% (HPLC)
Synonym(s):
(4E,8E)-5,9,13-Trimethyl-4,8,12-tetradecatrienoic acid (2E)-3,7-dimethyl-2,6-octadien-1-yl ester, (E,E,E)-4,8,12-Tetradecatrienoic acid, 5,9,13-trimetyl-, 3,7-dimetyl-2,6-octadienyl ester, Geranyl farnesylacetate
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
liquid
storage condition
desiccated
color
colorless to beige
storage temp.
−20°C
SMILES string
CC(C)=CCC/C(C)=C/COC(CC/C=C(CC/C=C(CCC=C(C)C)\C)\C)=O
InChI
1S/C27H44O2/c1-22(2)12-8-14-24(5)16-10-17-25(6)18-11-19-27(28)29-21-20-26(7)15-9-13-23(3)4/h12-13,16,18,20H,8-11,14-15,17,19,21H2,1-7H3/b24-16+,25-18+,26-20+
InChI key
ZPACYDRSPFRDHO-ROBAGEODSA-N
Biochem/physiol Actions
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service