Skip to Content
Merck
All Photos(1)

Key Documents

SML3182

Sigma-Aldrich

Eltrombopag

≥98% (HPLC)

Synonym(s):

(Z)-3′-(2-(1-(3,4-Dimethylphenyl)-3-methyl-5-oxo-1H-pyrazol-4(5H)-ylidene)hydrazinyl)-2′-hydroxybiphenyl-3-carboxylic acid, 3′-[(2Z)-2-[1-(3,4-Dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2′-hydroxy[1,1′-biphenyl]-3-carboxylic acid, SB 497115, SB-497115, SB497115

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$777.00
50 MG
R$3,142.00

R$777.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$777.00
50 MG
R$3,142.00

About This Item

Empirical Formula (Hill Notation):
C25H22N4O4
CAS Number:
Molecular Weight:
442.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$777.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(C=CC=C1N/N=C2C(C)=NN(C/2=O)C3=CC(C)=C(C=C3)C)C4=CC=CC(C(O)=O)=C4

Biochem/physiol Actions

Eltrombopag is an orally active nonpeptide thrombopoietin receptor (TpoR) agonist that selectively induces STAT activation and proliferation (EC50 = 270 & 30 nM, respectively) of hToR-transfected, but not non-transfected, BAF3 cells. Eltrombopag increases the differentiation of primary human CD34(+) bone marrow cells into CD41(+) megakaryocytes ex vivo (EC50 = 100 nM) and exhibits thrombopoietic activity in chimpanzees in vivo (10 mg/kg/day p.o.).
Orally active, potent and selective thrombopoietin (Tpo) receptor (TpoR) agonist in vitro and in vivo.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service