Skip to Content
Merck
All Photos(1)

Key Documents

SML3168

Sigma-Aldrich

VU0469650 hydrochloride

≥98% (HPLC)

Synonym(s):

VU0469650 hydrochloride, 3-[(3R)-3-Methyl-4-(tricyclo[3.3.1.13,7]dec-1-ylcarbonyl)-1-piperazinyl]-2-pyridinecarbonitrile, hydrochloride salt, VU 0469650 HCl, VU-0469650 HCl

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$1,393.00
50 MG
R$5,605.00

R$1,393.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$1,393.00
50 MG
R$5,605.00

About This Item

Empirical Formula (Hill Notation):
C22H28N4O · HCl
Molecular Weight:
400.94
UNSPSC Code:
12352200
NACRES:
NA.77

R$1,393.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -41 to -51°, c = 1 in methanol

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Brain-penetrant, potent and selective metabotropic glutamate receptor 1 (mGlu1) negative allosteric modulator (NAM).
VU0469650 is a brain-penetrant, potent and selective metabotropic glutamate receptor 1 (mGlu1) negative allosteric modulator (NAM). VU0469650 selectively inhibits glu-induced Ca2+ influx in human mGluR1-expressing HEK 293A (EC50 = 99 nM with [Glu] = EC20 for 1.9 min, then EC100 for 1.7 min; inactive against cells expressing human mGlu2-8 at 10 μM) with no affinity toward a panel of 68 clinically relevant GPCRs, ion channels, kinases, and transporters. Prior administration of VU0469650 in mice in vivo (60 mg/kg i.p.) modulates the antipsychotic activity of M4 muscarinic receptor PAM VU0467154 and that of the M1/M4 agonist xanomeline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service