Skip to Content
Merck
All Photos(1)

Key Documents

SML3164

Sigma-Aldrich

CAM833

≥98% (HPLC)

Synonym(s):

CAM833A, N-(2-((2S,4R)-2-(((S)-1-(2-chloro-4-methoxyphenyl)ethyl)carbamoyl)-4-hydroxypyrrolidin-1-yl)-2-oxoethyl)-6-fluoroquinoline-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$986.00
25 MG
R$3,960.00

R$986.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$986.00
25 MG
R$3,960.00

About This Item

Empirical Formula (Hill Notation):
C26H26ClFN4O5
Molecular Weight:
528.96
UNSPSC Code:
12352107
NACRES:
NA.77

R$986.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(N=C(C(NCC(N2[C@H](C(N[C@H](C3=C(Cl)C=C(OC)C=C3)C)=O)C[C@@H](O)C2)=O)=O)C=C4)C4=C1

InChI key

PMUWBFKMLGLUTF-KNUWZQJKSA-N

Biochem/physiol Actions

CAM833 is a potent and selective inhibitor of RAD51:BRCA2 interaction that inhibits RAD51 foci formation in A549 non-small cell lung carcinoma (NSCLC) cells. CAM833 suppresses extended RAD51 filament assembly. It potentiates the DNA damage by ionizing radiation in cells.
potent and selective inhibitor of RAD51:BRCA2 interaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duncan E Scott et al.
Cell chemical biology, 28(6), 835-847 (2021-03-05)
BRCA2 controls RAD51 recombinase during homologous DNA recombination (HDR) through eight evolutionarily conserved BRC repeats, which individually engage RAD51 via the motif Phe-x-x-Ala. Using structure-guided molecular design, templated on a monomeric thermostable chimera between human RAD51 and archaeal RadA, we
Duncan E Scott et al.
Cell chemical biology, 28(6), 835-847 (2021-03-05)
BRCA2 controls RAD51 recombinase during homologous DNA recombination (HDR) through eight evolutionarily conserved BRC repeats, which individually engage RAD51 via the motif Phe-x-x-Ala. Using structure-guided molecular design, templated on a monomeric thermostable chimera between human RAD51 and archaeal RadA, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service