Skip to Content
Merck
All Photos(1)

Key Documents

SML3151

Sigma-Aldrich

PF-06819784

≥98% (HPLC)

Synonym(s):

(2R,3R)-1-[[3-[4-[2-(4-Morpholinyl)-5-pyrimidinyl]phenyl]-1H-1,2,4-triazol-1-yl]carbonyl]-2-phenyl-3-piperidinecarboxylic acid, C11, LYPLAL1 inhibitor C11, LYPLAL1-IN-11

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$554.00
25 MG
R$2,234.00

R$554.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$554.00
25 MG
R$2,234.00

About This Item

Empirical Formula (Hill Notation):
C29H29N7O4
CAS Number:
Molecular Weight:
539.59
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

R$554.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

C11 (LYPLAL1-IN-11) is a cell penetrant potent and selective covalent inhibitor of lysophospholipase-like 1 (LYPLAL1). C11 significant increase in glucose production in human, rat, and mouse hepatocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service