Skip to Content
Merck
All Photos(1)

Key Documents

SML3074

Sigma-Aldrich

Gap26 trifluoroacetate

≥95% (HPLC)

Synonym(s):

Gap 26 trifluoroacetate, Gap-26 trifluoroacetate, L-Arginine, L-valyl-L-cysteinyl-L-tyrosyl-L-?-aspartyl-L-lysyl-L-seryl-L-phenylalanyl-L-prolyl-L-isoleucyl-L-seryl-L-histidyl-L-valyl-, trifluoroacetate, VCYDKSFPISHVR trifluoroacetate, Val-Cys-Tyr-Asp-Lys-Ser-Phe-Pro-Ile-Ser-His-Val-Arg trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$2,401.00

R$2,401.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$2,401.00

About This Item

Empirical Formula (Hill Notation):
C70H107N19O19S · xC2HF3O2
CAS Number:
Molecular Weight:
1550.78 (free base basis)
UNSPSC Code:
41106609
NACRES:
NA.77

R$2,401.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

film or powder

color

white to off-white

storage temp.

−20°C

SMILES string

SC[C@H](NC(=O)[C@@H](N)C(C)C)C(=O)N[C@@H](Cc4ccc(cc4)O)C(=O)N[C@@H](CC(=O)O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccccc3)C(=O)N1[C@@H](CCC1)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc2nc[nH]c2)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCNC

InChI

1S/C70H107N19O19S/c1-7-38(6)56(67(105)85-50(33-91)61(99)81-46(29-41-31-75-35-77-41)60(98)87-55(37(4)5)66(104)79-44(69(107)108)18-13-25-76-70(73)74)88-64(102)52-19-14-26-89(52)68(106)48(28-39-15-9-8-10-16-39)83-62(100)49(32-90)84-57(95)43(17-11-12-24-71)78-59(97)47(30-53(93)94)82-58(96)45(27-40-20-22-42(92)23-21-40)80-63(101)51(34-109)86-65(103)54(72)36(2)3/h8-10,15-16,20-23,31,35-38,43-52,54-56,90-92,109H,7,11-14,17-19,24-30,32-34,71-72H2,1-6H3,(H,75,77)(H,78,97)(H,79,104)(H,80,101)(H,81,99)(H,82,96)(H,83,100)(H,84,95)(H,85,105)(H,86,103)(H,87,98)(H,88,102)(H,93,94)(H,107,108)(H4,73,74,76)/t38-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,54-,55-,56-/m0/s1

InChI key

FDPIMWZHGJNESB-VCSXYVMHSA-N

Biochem/physiol Actions

Connexin-43 (Cx43) etracellular loop I sequence-derived Cx43 blocker in cultures and in animals in vivo.
Gap 26 is a 13-mer peptide derived from the first extracellular loop I sequence of connexin-43 (h/m/r Cx43 aa64-76 or aa63-75 after Met1 removal). Gap 26 is widely employed to block and probe Cx43-mediated gap junctional signaling in cultures (20-160 μM) and in animals in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service