Skip to Content
Merck
All Photos(1)

Key Documents

SML3019

Sigma-Aldrich

TH1760

≥98% (HPLC)

Synonym(s):

6-{[4-(1H-indole-5-carbonyl)piperazin-1-yl]sulfonyl}-2,3-dihydro-1,3-benzoxazol-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$777.00
25 MG
R$3,142.00

R$777.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$777.00
25 MG
R$3,142.00

About This Item

Empirical Formula (Hill Notation):
C20H18N4O5S
CAS Number:
Molecular Weight:
426.45
UNSPSC Code:
12352200
NACRES:
NA.77

R$777.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC(O2)=C(C=C1)NC2=O)(N3CCN(CC3)C(C4=CC5=C(C=C4)NC=C5)=O)=O

Biochem/physiol Actions

TH1760 is a cell penetrant, potent and selective inhibitor of NUDT15 (NUDIX-type 15) that directly binds into the NUDT15 catalytic pocket. TH1760 sensitized cells to 6-thioguanine.
cell penetrant, potent and selective inhibitor of NUDT15 (NUDIX-type 15) that directly binds into the NUDT15 catalytic pocket

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Si Min Zhang et al.
Nature chemical biology, 16(10), 1120-1128 (2020-07-22)
The NUDIX hydrolase NUDT15 was originally implicated in sanitizing oxidized nucleotides, but was later shown to hydrolyze the active thiopurine metabolites, 6-thio-(d)GTP, thereby dictating the clinical response of this standard-of-care treatment for leukemia and inflammatory diseases. Nonetheless, its physiological roles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service