Skip to Content
Merck
All Photos(1)

Key Documents

SML2865

Sigma-Aldrich

D5D-IN-326

≥98% (HPLC)

Synonym(s):

2-(2,2,3,3,3-Pentafluoropropoxy)-3-[4-(2,2,2-trifluoroethoxy)phenyl]-5,7-dihydro-3H-pyrrolo[2,3-d]pyrimidine-4,6-dione, Compound-326

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$933.00
25 MG
R$3,751.00

R$933.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$933.00
25 MG
R$3,751.00

About This Item

Empirical Formula (Hill Notation):
C17H11F8N3O4
CAS Number:
Molecular Weight:
473.27
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

R$933.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)COc1[n]([c](c3c([nH]c(c3)O)n1)=O)c2ccc(cc2)OCC(F)(F)F

InChI

1S/C17H11F8N3O4/c18-15(19,17(23,24)25)6-32-14-27-12-10(5-11(29)26-12)13(30)28(14)8-1-3-9(4-2-8)31-7-16(20,21)22/h1-5,26,29H,6-7H2

InChI key

VKDOANVHEBFXOC-UHFFFAOYSA-N

Biochem/physiol Actions

D5D-IN-326 (Compound-326) is an orally available, potent and selective inhibitor of delta-5 desaturase (D5D) that reduces insulin resistance and body weight in a high-fat diet-induced obese mice. It appears that D5D-IN-326 significantly inhibits the progression of atherosclerosis in the aorta of western-diet fed ApoE knockout (KO) mice. It is called Compound-326 in recent J Pharmacol Exp Ther. 2019 Nov;371(2):290-298. (PMID: 31488602) paper.
orally available, potent and selective inhibitor of delta-5 desaturase (D5D)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuichi Takagahara et al.
The Journal of pharmacology and experimental therapeutics, 371(2), 290-298 (2019-09-07)
Delta-5 desaturase (D5D), encoded by fatty acid desaturase 1 (Fads1), is the rate-limiting enzyme for the conversion from dihomo-γ-linolenic acid (DGLA) to arachidonic acid (AA) in the ω-6 polyunsaturated fatty acid pathway. Several AA-derived eicosanoids (e.g., prostaglandins, thromboxanes, and leukotrienes)
Hiroaki Yashiro et al.
PloS one, 11(11), e0166198-e0166198 (2016-11-11)
Obesity is now recognized as a state of chronic low-grade inflammation and is called as metabolic inflammation. Delta-5 desaturase (D5D) is an enzyme that metabolizes dihomo-γ-linolenic acid (DGLA) to arachidonic acid (AA). Thus, D5D inhibition increases DGLA (precursor to anti-inflammatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service