Skip to Content
Merck
All Photos(1)

Key Documents

SML2748

Sigma-Aldrich

BAY-850 trihydrochloride

≥98% (HPLC)

Synonym(s):

N-((R)-1-((1s,4S)-4-Aminocyclohexylamino)-3-(4-cyanophenyl)propan-2-yl)-2-chloro-4-methoxy-5-(5-(((R)-1-p-tolylethylamino)methyl)furan-2-yl)benzamide trihydrochloride, N-[(2R)-1-[(4-Aminocyclohexyl)amino]-3-(4-cyanophenyl)propan-2-yl]-2-chloro-4-methoxy-5-[5-({[(1R)-1-(4-methylphenyl)ethyl]amino}methyl)-2-furyl]benzamide trihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$2,112.00
25 MG
R$8,016.00

R$2,112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$2,112.00
25 MG
R$8,016.00

About This Item

Empirical Formula (Hill Notation):
C38H44ClN5O3 · 3HCl
CAS Number:
Molecular Weight:
763.62
UNSPSC Code:
12352200
NACRES:
NA.77

R$2,112.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

BAY-850 is a chemical probe that targets ATPase family AAA domain-containing protein 2 (ATAD2) bromodomain (BD) with submicromolar affinity (Kd of 84.9 nM by MST and 120 nM by BROMOscan) and high selectivity over all other bromodomains tested (no BROMOscan hits at 10 μM). BAY-850 blocks ATAD2 BD from binding acetylated H4 peptide in cell-free assays (IC50 of 20 nM) and displaces full-length ATAD2 from chromatin in live cells (effctive conc. 1 μM by HTRF). To avoid potential unspecific off-target effects, concentrations above 5 μM are not recommended. BAY-460 is a companion control compound with strongly reduced ATAD2 inhibitory potency (IC50 = 16 μM by cell-based HTRF assay; no activity at 10 μM by BROMOscan).
Chemical probe that targets ATAD2 bromodomain (BD) with submicromolar affinity and displaces full-length ATAD2 from chromatin in live cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service