Skip to Content
Merck
All Photos(1)

Key Documents

SML2716

Sigma-Aldrich

Pitolisant hydrochloride

≥98% (HPLC)

Synonym(s):

1-[3-[3-(4-Chlorophenyl)propoxy]propyl]piperidine hudrochloride, 1-{3-[3-(4-Chlorophenyl)propoxy]propyl}piperidine hydrochloride, 3-(4-Chlorophenyl)propyl 3-piperidinopropyl ether hydrochloride, BF 2649, BF2.649, Ciproxidine, Tiprolisant

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$741.00
50 MG
R$3,007.00

R$741.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$741.00
50 MG
R$3,007.00

About This Item

Empirical Formula (Hill Notation):
C17H26ClNO · HCl
CAS Number:
Molecular Weight:
332.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$741.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)CCCOCCCN2CCCCC2.Cl

InChI

1S/C17H26ClNO.ClH/c18-17-9-7-16(8-10-17)6-4-14-20-15-5-13-19-11-2-1-3-12-19;/h7-10H,1-6,11-15H2;1H

InChI key

XLFKECRRMPOAQS-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective competitive antagonist/inverse agonist of histamine H3 receptor

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service