Skip to Content
Merck
All Photos(1)

Key Documents

SML2686

Sigma-Aldrich

THAL-SNS-032

≥98% (HPLC)

Synonym(s):

4-[[[5-[[[5-(1,1-Dimethylethyl)-2-oxazolyl]methyl]thio]-2-thiazolyl]amino]carbonyl]-N-[2-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]ethoxy]ethoxy]ethoxy]ethyl]-1-piperidineacetamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$837.00
5 MG
R$3,187.00

R$837.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
R$837.00
5 MG
R$3,187.00

About This Item

Empirical Formula (Hill Notation):
C40H52N8O10S2
CAS Number:
Molecular Weight:
869.02
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$837.00


Check Cart for Availability

Request a Bulk Order

ligand

pomalidomide

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c(ncc1SCc6ncc([o]6)C(C)(C)C)NC(=O)C2CCN(CC2)CC(=O)NCCOCCOCCOCCNc3c4c(ccc3)C(=O)N(C4=O)C5CCC(=O)NC5=O

InChI

1S/C40H52N8O10S2/c1-40(2,3)29-21-43-32(58-29)24-59-33-22-44-39(60-33)46-35(51)25-9-13-47(14-10-25)23-31(50)42-12-16-56-18-20-57-19-17-55-15-11-41-27-6-4-5-26-34(27)38(54)48(37(26)53)28-7-8-30(49)45-36(28)52/h4-6,21-22,25,28,41H,7-20,23-24H2,1-3H3,(H,42,50

InChI key

BXDZOYLPNAIDOC-UHFFFAOYSA-N

Biochem/physiol Actions

Selective inhibitor and degrader of CDK9
THAL-SNS-032 is a selective inhibitor and degrader of cyclin-dependent kinase 9 (CDK9). It links the CDK inhibitor SNS-032 to a thalidomide derivative that binds the E3 ubiquitin ligase Cereblon. THAL-SNS-032 was found to selectively induce degradation of CDK9 over other CDKs. It inhibited proliferation across a panel of 11 different leukemia cancer cell lines potently than SNS-032.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calla M Olson et al.
Nature chemical biology, 14(2), 163-170 (2017-12-19)
Cyclin-dependent kinase 9 (CDK9), an important regulator of transcriptional elongation, is a promising target for cancer therapy, particularly for cancers driven by transcriptional dysregulation. We characterized NVP-2, a selective ATP-competitive CDK9 inhibitor, and THAL-SNS-032, a selective CDK9 degrader consisting of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service