Skip to Content
Merck
All Photos(1)

Key Documents

SML2670

Sigma-Aldrich

BAY-8002

≥98% (HPLC)

Synonym(s):

2-(2-chloro-5-(phenylsulfonyl)benzamido)benzoic acid, 2-[[2-Chloro-5-(phenylsulfonyl)benzoyl]amino]-benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$789.00
25 MG
R$3,188.00

R$789.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$789.00
25 MG
R$3,188.00

About This Item

Empirical Formula (Hill Notation):
C20H14ClNO5S
CAS Number:
Molecular Weight:
415.85
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$789.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(c2cc(c(cc2)Cl)C(=O)Nc3c(cccc3)C(=O)O)c1ccccc1

InChI

1S/C20H14ClNO5S/c21-17-11-10-14(28(26,27)13-6-2-1-3-7-13)12-16(17)19(23)22-18-9-5-4-8-15(18)20(24)25/h1-12H,(H,22,23)(H,24,25)

InChI key

CLAUJSRBKSRTGQ-UHFFFAOYSA-N

Biochem/physiol Actions

BAY-8002 is an orally available, potent and selective inhibitor of lactate transporter MCT1 (monocarboxylate transporter 1; SLC16A1) that potently inhibits bidirectional lactate transport. BAY-8002 exhibits antiproliferative activity in numerous cancer cell lines. BAY-8002 induces accumulation of lactate and significantly inhibits tumor growth in mice model. It does not cause tumor regression.
orally available, potent and selective inhibitor of lactate transporter MCT1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Quanz et al.
Molecular cancer therapeutics, 17(11), 2285-2296 (2018-08-18)
The lactate transporter SLC16A1/monocarboxylate transporter 1 (MCT1) plays a central role in tumor cell energy homeostasis. In a cell-based screen, we identified a novel class of MCT1 inhibitors, including BAY-8002, which potently suppress bidirectional lactate transport. We investigated the antiproliferative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service