Skip to Content
Merck
All Photos(1)

Key Documents

SML2649

Sigma-Aldrich

T3-CLK-N

≥98% (HPLC)

Synonym(s):

N-(6-(3,5-Di-tert-butylphenyl)imidazo[1,2-a]pyridin-2-yl)-4-(2-methyl-1-(4-methylpiperazin-1-yl)-1-oxopropan-2-yl)benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$631.00
25 MG
R$2,543.00

R$631.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$631.00
25 MG
R$2,543.00

About This Item

Empirical Formula (Hill Notation):
C37H47N5O2
Molecular Weight:
593.80
UNSPSC Code:
12352200
NACRES:
NA.77

R$631.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

T3-CLK-N (T3N) is a negative control for CDC-like kinase (CLK) inhibitor T3-CLK.
negative control for CDC-like kinase (CLK) inhibitor T3-CLK

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler Funnell et al.
Nature communications, 8(1), 7-7 (2017-02-25)
CDC-like kinase phosphorylation of serine/arginine-rich proteins is central to RNA splicing reactions. Yet, the genomic network of CDC-like kinase-dependent RNA processing events remains poorly defined. Here, we explore the connectivity of genomic CDC-like kinase splicing functions by applying graduated, short-exposure
Susanne Müller et al.
eLife, 7 (2018-04-21)
Potent, selective and broadly characterized small molecule modulators of protein function (chemical probes) are powerful research reagents. The pharmaceutical industry has generated many high-quality chemical probes and several of these have been made available to academia. However, probe-associated data and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service