Skip to Content
Merck
All Photos(1)

Key Documents

SML2625

Sigma-Aldrich

RCM-1

≥98% (HPLC)

Synonym(s):

2-[[2-Oxo-2-(2-thienyl)ethyl]thio]-4,6-di-2-thienyl-3-pyridinecarbonitrile, 2-{[2-Oxo-2-(thiophen-2-yl)ethyl]sulfanyl}-4,6-di(thiophen-2-yl)pyridine-3-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$670.00
25 MG
R$2,698.00

R$670.00


Estimated to ship onApril 23, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$670.00
25 MG
R$2,698.00

About This Item

Empirical Formula (Hill Notation):
C20H12N2OS4
CAS Number:
Molecular Weight:
424.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$670.00


Estimated to ship onApril 23, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to very dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(ccc1)c2nc(c(c(c2)c4[s]ccc4)C#N)SCC(=O)c3[s]ccc3

InChI key

DCCYTKKDAZEVLY-UHFFFAOYSA-N

Biochem/physiol Actions

Induces FOXM1 degradation and blocks its nuclear localization, suppresses FOXM1-mediated goblet cell metaplasia and airway hyperresponsiveness in vivo.
RCM-1 is a forkhead box M1 (FOXM1) inhibitor that blocks FOXM1 nuclear localization (IC50 = 720 nM using GFP-FOXM1-expressing U2OS cells) and causes FOXM1 degradation via ubiquitination induction (2.9- and 4.8-fold of basal level in A549 cells with 10 μM or 20 μM RCM-1, respectively) without affecting cellular YAP, FACT140, NF-κB, FOXA2, and FOXJ1 levels. RCM-1 intraperitoneal injection (1.7 mg/kg) in mice downregulates endogenous murine FOXM1 level as well as transgenic human FOXM1 GFP fusion level in vivo, effectively suppressing house dust mite-induced lung inflammation as well as goblet cell metaplasia and airway hyperresponsiveness in response to IL-13.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaohui Wu et al.
Biochemical and biophysical research communications, 495(1), 1432-1439 (2017-11-22)
Functional abnormal airway epithelial cells, along with activated inflammatory cells, resulting in chronic airway inflammation, are considered as the characteristic of asthma. Fatty Acid Binding Protein 4 (FABP4) takes part in glucose and lipid homeostasis, and also have an important
Lifeng Sun et al.
Science signaling, 10(475) (2017-04-20)
Goblet cell metaplasia and excessive mucus secretion associated with asthma, cystic fibrosis, and chronic obstructive pulmonary disease contribute to morbidity and mortality worldwide. We performed a high-throughput screen to identify small molecules targeting a transcriptional network critical for the differentiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service