FRAX1036 is a potent, group I-selective p21-activated kinase (PAK) inhibitor (PAK1/4 Ki = 23 nM/1.4 μM; PAK1/4 IC50 = 2 nM/>500 nM; >96% PAK1/2/3 inhibition vs. 5.1%/8.2%/22.2% PAK4/5/6 inhibition at 1 μM) with significant inhibitory activity toward only 11 other kinases at a high concentration of 1 μM (78.7-100% inhibition) among a panel of 240. FRAX1036 exhibits higher antiproliferation activity in PAK negative regulator NF2-deficient meningioma cultures (NF2-/- Ben-Men-1/KT21-MG1 IC50 = 1.888/1.991 μM vs.NF2+/+ MN328/MN525 IC50 = 4.239/3.296 μM) by inhibiting cellular PAK autophosphorylation and downstream signaling events. When administered orally in a murine orthotopic meningioma model, FRAX1036 effectively suppresses the growth of established KT21 and Ben-Men tumors in vivo (30 mg/kg/d p.o.).
Orally active, potent group I-selective p21-activated kinase (PAK1/2/3) inhibitor with in vivo efficacy in a murine orthotopic meningioma model.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Malignant peripheral nerve sheath tumors (MPNSTs) are devastating sarcomas for which no effective medical therapies are available. Over 50% of MPSNTs are associated with mutations in NF1 tumor suppressor gene, resulting in activation of Ras and its effectors, including the
Molecular and cellular biology, 35(23), 3990-4005 (2015-09-24)
p21-activated kinases (Paks) have been shown to regulate cytoskeleton rearrangements, cell proliferation, attachment, and migration in a variety of cellular contexts, including endothelial cells. However, the role of endothelial Pak in embryo development has not been reported, and currently, there
The p21-activated kinases (PAKs) are Cdc42/Rac-activated serine-threonine protein kinases that regulate several key cancer-relevant signaling pathways, such as the Mek/Erk, PI3K/Akt, and Wnt/β-catenin cascades. Pak1 is frequently overexpressed and/or hyperactivated in different human cancers, including breast, ovary, prostate, and brain
Journal of medicinal chemistry, 58(1), 111-129 (2014-11-22)
The p21-activated kinase (PAK) family of serine/threonine protein kinases plays important roles in cytoskeletal organization, cellular morphogenesis, and survival, and members of this family have been implicated in many diseases including cancer, infectious diseases, and neurological disorders. Owing to their
Megakaryocyte maturation and polyploidization are critical for platelet production; abnormalities in these processes are associated with myeloproliferative disorders, including thrombocytopenia. Megakaryocyte maturation signals through cascades that involve p21-activated kinase (Pak) function; however, the specific role for Pak kinases in megakaryocyte
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.