Skip to Content
Merck
All Photos(1)

Key Documents

SML2579

Sigma-Aldrich

Tafluprost acid

≥98% (HPLC)

Synonym(s):

(5Z)-7-[(1R,2R,3R,5S)-2-[(1E)-3,3-difluoro-4-phenoxy-1-buten-1-yl]-3,5-dihydroxycyclopentyl]-5-Heptenoic acid, 9α,11α-dihydroxy-15,15-difluoro-16-phenoxy-17,18,19,20-tetranor-prosta-5Z,13E-dien-1-oic acid, AFP-172

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$473.00
5 MG
R$1,915.00

R$473.00


Check Cart for Availability


Select a Size

Change View
1 MG
R$473.00
5 MG
R$1,915.00

About This Item

Empirical Formula (Hill Notation):
C22H28F2O5
CAS Number:
Molecular Weight:
410.45
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

R$473.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

oil

optical activity

[α]/D 17 to 22°, c = 1.0 in methanol

color

colorless

storage temp.

−20°C

SMILES string

FC(F)(COc2ccccc2)\C=C\[C@@H]1[C@H]([C@H](C[C@H]1O)O)C\C=C/CCCC(=O)O

InChI

1S/C22H28F2O5/c23-22(24,15-29-16-8-4-3-5-9-16)13-12-18-17(19(25)14-20(18)26)10-6-1-2-7-11-21(27)28/h1,3-6,8-9,12-13,17-20,25-26H,2,7,10-11,14-15H2,(H,27,28)/b6-1-,13-12+/t17-,18-,19+,20-/m1/s1

InChI key

KIQXRQVVYTYYAZ-VKVYFNERSA-N

Biochem/physiol Actions

Prostaglandin F2α agonist; Antiglaucoma agent
Taflupost acid is the active metabolite of Tafluprost, a prostaglandin F2α agonist that lowers intraocular pressure and is used in the treatment of glaucoma. Taflupost acid has high affinity and selectivity for the prostaglandin PGF2α (FP) receptor with a Ki of 0.4 nM.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service