Skip to Content
Merck
All Photos(1)

Key Documents

SML2571

Sigma-Aldrich

CPHPC

≥98% (HPLC)

Synonym(s):

(2′R)-Adipoyldi-D-proline, (R)-1-[6-[(R)-2-Carboxy-pyrrolidin-1-yl]-6-oxo-hexanoyl-]pyrrolidine-2-carboxylic acid, 1,1′-(1,6-Dioxo-1,6-hexanediyl)bis[D-proline], GSK 2315698, GSK-2315698, GSK2315698, Miridesap, R-1-[6-[R-2-carboxy-pyrrolidin-1-yl]-6-oxohexanoyl]pyrrolidine-2-carboxylic acid, Ro 63-8695, Ro-63-8695

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$426.00
25 MG
R$1,734.00

R$426.00


Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$426.00
25 MG
R$1,734.00

About This Item

Empirical Formula (Hill Notation):
C16H24N2O6
CAS Number:
Molecular Weight:
340.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$426.00


Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N2([C@H](CCC2)C(=O)O)C(=O)CCCCC(=O)N1[C@H](CCC1)C(=O)O

InChI

1S/C16H24N2O6/c19-13(17-9-3-5-11(17)15(21)22)7-1-2-8-14(20)18-10-4-6-12(18)16(23)24/h11-12H,1-10H2,(H,21,22)(H,23,24)/t11-,12-/m1/s1

InChI key

HZLAWYIBLZNRFZ-VXGBXAGGSA-N

Biochem/physiol Actions

CPHPC (Miridesap; Ro 63-8695) is an orally active serum amyloid P (SAP) blocker that targets the ligand-binding site on the SAP planar binding (B) face with each of its two D-proline moieties (up to two SAP per CPHPC), being able to crosslink and dimerize two SAP pentamers to form B face-to-B face decamers. CPHPC potently inhibits SAP binding to amyloid fibrils in vitro (IC50 = 0.9 μM using AD Aβ amyloid fibrils) and exhibits in vivo efficacy toward liver & circulating SAP clearance as well as SAP removal from tissue amyloid deposits (1-5 mg/mL in drinking water; human SAP transgenic mice).
Serum amyloid P (SAP) blocker that inhibits SAP binding to amyloid fibrils, clears circulating SAP and removes SAP from tissue amyloid deposits in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service