Skip to Content
Merck
All Photos(1)

Key Documents

SML2563

Sigma-Aldrich

TFMB-(S)-2HG

≥98% (HPLC)

Synonym(s):

(2S)-Tetrahydro-5-oxo-2-furancarboxylic acid, [3-(trifluoromethyl)phenyl]methyl ester, (S)-2-HG-TFMB, (S)-2HG-TFMB, (S)-3-(Trifluoromethyl)benzyl 5-oxotetrahydrofuran-2-carboxylate, (S)-TFMB-2-HG, (S)-TFMB-2HG, (S)-TFMB-OTC, TFMB-(S)-2-HG, TFMB-L2HG, Trifluoromethylbenzyl-(S)-2-HG

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$807.00

R$807.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$807.00

About This Item

Empirical Formula (Hill Notation):
C13H11F3O4
CAS Number:
Molecular Weight:
288.22
UNSPSC Code:
12352200
NACRES:
NA.77

R$807.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)(F)c1cc(ccc1)COC(=O)[C@H]2OC(=O)CC2

InChI key

UXTLNUGKDZMPTC-JTQLQIEISA-N

Biochem/physiol Actions

Membrane-permeant ester precursor of (S)-2-hydroxyglutarate (L-2HG) with distinct cellular activity as the oncometabolite (R)-2HG (D-2HG).
TFMB-(S)-2HG is a membrane-permeant (S)-2-hydroxyglutarate (L-2HG) precursor used in conjunction with the oncometabolite (R)-2-hydroxyglutarate (D-2HG) precursor for probing 2-HG enantiomer-specific cellular responses (typical conc used in culture: 0.5-20 mM). While both 2HG enantiomers inhibit multiple α-ketoglutarate (α-KG; 2-OG)-dependent dioxygenases by competing against α-KG binding, (S)-2HG is reported to be more potent against TET1/2, CeKDM7A & KDM5B (JARID1B/PLU-1). On the other hand, (R)-2HG, but not (S)-2HG, is shown to downregulate HIF-1a by promoting EglN1 (HIF prolyl hydroxylase 2; HIF-PH2; PHD2) activity.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service