Skip to Content
Merck
All Photos(1)

Key Documents

SML2534

Sigma-Aldrich

DJ-V-159

≥98% (HPLC)

Synonym(s):

N1,N3-Bis(4-cyano-3-(trifluoromethyl)phenyl)isophthalamide, N1,N3-bis[4-cyano-3-(trifluoromethyl)phenyl]-1,3-Benzenedicarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$608.00
25 MG
R$2,460.00

R$608.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$608.00
25 MG
R$2,460.00

About This Item

Empirical Formula (Hill Notation):
C24H12F6N4O2
CAS Number:
Molecular Weight:
502.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$608.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC(C(NC2=CC=C(C#N)C(C(F)(F)F)=C2)=O)=CC=C1)NC3=CC(C(F)(F)F)=C(C#N)C=C3

InChI key

VEVNLJLJLVESLL-UHFFFAOYSA-N

Biochem/physiol Actions

DJ-V-159 is an agonist of GPRC6A, a family C G protein coupled receptor proposed to be master regulator of energy metabolism. GPRC6A is the receptor for osteocalcin, basic amino acids (e.g., L-Arginine), cations (e.g., calcium), and is a membrane receptor for testosterone. DJ-V-159 activated GPRC6A at 0.2 nM concentrations and stimulated GPRC6A-mediated ERK phosphorylation and cAMP accumulation in GPRC6A expressing HEK-293 cells. DJ-V-159 was found to stimulate insulin secretion from mouse β-cells and and lower serum glucose in wild-type mice.
GPRC6A agonist found to stimulate insulin secretion from mouse β-cells and and lower serum glucose in wild-type mice

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service