Skip to Content
Merck
All Photos(1)

Key Documents

SML2531

Sigma-Aldrich

TH5487

≥98% (HPLC)

Synonym(s):

4-(4-Bromo-2-oxo-3H-benzimidazol-1-yl)-N-(4-iodophenyl)piperidine-1-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,316.00
25 MG
R$5,096.00

R$1,316.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$1,316.00
25 MG
R$5,096.00

About This Item

Empirical Formula (Hill Notation):
C19H18BrIN4O2
CAS Number:
Molecular Weight:
541.18
UNSPSC Code:
12352200
NACRES:
NA.77

R$1,316.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

IC1=CC=C(NC(N2CCC(N(C3=CC=CC(Br)=C3N4)C4=O)CC2)=O)C=C1

Related Categories

Biochem/physiol Actions

Inhibitor of 8-oxoguanine DNA glycosylase 1 (OGG1)
TH5487 is an inhibitor of 8-oxoguanine DNA glycosylase 1 (OGG1), which is part of the DNA base excision repair pathway. TH5487 as shown to inhibit OGG1-dependent proinflammatory gene expression by binding in the OGG1 active site, preventing OGG1 from engaging damaged DNA in vitro and in cells, resulting in decreased proinflammatory gene expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Torkild Visnes et al.
Science (New York, N.Y.), 362(6416), 834-839 (2018-11-18)
The onset of inflammation is associated with reactive oxygen species and oxidative damage to macromolecules like 7,8-dihydro-8-oxoguanine (8-oxoG) in DNA. Because 8-oxoguanine DNA glycosylase 1 (OGG1) binds 8-oxoG and because Ogg1-deficient mice are resistant to acute and systemic inflammation, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service