Skip to Content
Merck
All Photos(1)

Key Documents

SML2520

Sigma-Aldrich

MI-503

≥98% (HPLC)

Synonym(s):

1-((1H-Pyrazol-4-yl)methyl)-4-methyl-5-((4-(6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-ylamino)piperidin-1-yl)methyl)-1H-indole-2-carbonitrile, 4-Methyl-1-(1H-pyrazol-4-ylmethyl)-5-[[4-[[6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl]amino]-1-piperidinyl]methyl]-1H-indole-2-carbonitrile, MI 503, MI503

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,008.00
25 MG
R$4,077.00

R$1,008.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$1,008.00
25 MG
R$4,077.00

About This Item

Empirical Formula (Hill Notation):
C28H27F3N8S
CAS Number:
Molecular Weight:
564.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$1,008.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)(F)Cc1[s]c2ncnc(c2c1)NC3CCN(CC3)Cc4c(c5c([n](c(c5)C#N)Cc6c[nH]nc6)cc4)C

InChI key

DETOMBLLEOZTMZ-UHFFFAOYSA-N

Biochem/physiol Actions

Highly potent menin-MLL interaction inhibitor that selectively inhibits MLL oncogenic fusions-driven cancer progression in vitro and in vivo.
MI-503 is a highly potent menin-MLL interaction inhibitor (cell-free IC50 = 14.7 nM against 1 nM FITC-MLL(4-13) peptide; cellular IC50 = 220 nM using HEK293 MLL-AF9 fusion transfectants) that targets menin MLL-binding site with high affinity (KD = 9.3 nM by ITC). MI-503 selectively inhibits mixed lineage leukemia (MLL) oncogenic fusion-driven cancer progression in cultures (GI50 from 250 to 570 nM; GI50 >5 μM against cells without MLL gene translocation) and is efficacious against human MLL leukemia MV4;11 tumor growth in mice in vivo (by 80-100%; 60 mg/kg/day i.p.) with good pharmacokinetic properties, oral bioavailability (73% post 30 mg/kg p.o.), and no adverse effects to the animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tasha L Lin et al.
Blood advances, 8(2), 261-275 (2023-12-04)
RNA-binding proteins (RBPs) are emerging as a novel class of therapeutic targets in cancer, including in leukemia, given their important role in posttranscriptional gene regulation, and have the unexplored potential to be combined with existing therapies. The RBP insulin-like growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service