Skip to Content
Merck
All Photos(1)

Key Documents

SML2336

Sigma-Aldrich

Sk052-145-2

≥97% (HPLC)

Synonym(s):

4-(Cyclopentyloxy)-5-methoxy-2-nitro-N-(5-(pentan-3-yl)-1,3,4-thiadiazol-2-yl)benzamide, Lipoprotein signal peptidase inhibitor, Lsp inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$695.00
25 MG
R$2,805.00

R$695.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$695.00
25 MG
R$2,805.00

About This Item

Empirical Formula (Hill Notation):
C20H26N4O5S
CAS Number:
Molecular Weight:
434.51
UNSPSC Code:
12352200
NACRES:
NA.77

R$695.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=NN=C(C(CC)CC)S1)C2=CC(OC)=C(OC3CCCC3)C=C2[N+]([O-])=O

InChI

1S/C20H26N4O5S/c1-4-12(5-2)19-22-23-20(30-19)21-18(25)14-10-16(28-3)17(11-15(14)24(26)27)29-13-8-6-7-9-13/h10-13H,4-9H2,1-3H3,(H,21,23,25)

InChI key

QRUAZOLDUDHNBK-UHFFFAOYSA-N

Biochem/physiol Actions

Sk052-145-2 is a potent and selective inhibitor of bacterial Lipoprotein signal peptidase (Lsp). Sk052-145-2 potently inhibits growth of E. Coli in a presence of polymyxin B nonapeptide (PMBN).
potent and selective inhibitor of bacterial Lipoprotein signal peptidase (Lsp)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seiya Kitamura et al.
Cell chemical biology, 25(3), 301-308 (2018-01-18)
As resistance to antibiotics increases, the exploration of new targets and strategies to combat pathogenic bacteria becomes more urgent. Ideal protein targets are required for viability across many species, are unique to prokaryotes to limit effects on the host, and have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service