Skip to Content
Merck
All Photos(1)

Key Documents

SML2271

Sigma-Aldrich

ADX-47273

≥98% (HPLC)

Synonym(s):

(4-Fluorophenyl)[(3S)-3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]-1-piperidinyl]methanone, (S)-(4-Fluorophenyl)-{3-[3-(4-fluorophenyl)-[1,2,4]oxadiazol-5-yl]-piperidin-1-yl}-methanone, ADX 47273, ADX47273

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$393.00
25 MG
R$1,586.00

R$393.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$393.00
25 MG
R$1,586.00

About This Item

Empirical Formula (Hill Notation):
C20H17F2N3O2
CAS Number:
Molecular Weight:
369.36
MDL number:
UNSPSC Code:
12352200

R$393.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +90 to +110°, c = 0.1 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=CC=C(C=C1)C2=NOC([C@@H]3CN(CCC3)C(C4=CC=C(C=C4)F)=O)=N2

InChI

1S/C20H17F2N3O2/c21-16-7-3-13(4-8-16)18-23-19(27-24-18)15-2-1-11-25(12-15)20(26)14-5-9-17(22)10-6-14/h3-10,15H,1-2,11-12H2/t15-/m0/s1

InChI key

VXQCCZHCFBHTTD-HNNXBMFYSA-N

Biochem/physiol Actions

ADX-47273 is a brain-penetrating, potent and selective metabotropic glutamate receptor 5 (mGlu5; mGluR5) positive allosteric modulator (PAM) that enhances glutamate-stimulated Ca2+ response in rat cortical astrocytes (EC50 = 170 nM, Emax = 380%; [glutamate] = EC20 = 200 nM) via direct mGlu5 binding in a MPEP-, but not Quisqualate-, competitive manner (Ki = 4.3 μM against 2 nM MPEP; rat mGlu5 HEK293) with little mGlu5 agonist activity, no potency toward other rat/human family III GPCRs (mGlu1–8 and GABA-B), nor affinity to 56 GPCRs/transporters/enzymes/ion channels. ADX-47273 shows in vivo antipsychotic-like and procognitive efficacy in mice and rats in vivo (1-300 mg/kg i.p.) with good pharmacokinetic properties (B/P ratio >2, bioavailability ∼40%, T1/2 ∼2 hrs in rats).
Brain-penetrant, potent and selective mGlu5 (mGluR5) positive allosteric modulator (PAM) with antipsychotic and procognitive efficacy in mice and rats in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah N Isherwood et al.
Journal of neurochemistry, 145(2), 111-124 (2018-01-10)
Dysregulation of prefrontal cortical glutamatergic signalling via NMDA receptor hypofunction has been implicated in cognitive dysfunction and impaired inhibitory control in such neuropsychiatric disorders as schizophrenia, attention-deficit hyperactivity disorder and drug addiction. Although NMDA receptors functionally interact with metabotropic glutamate
Arthur Bikbaev et al.
Neuropharmacology, 115, 20-29 (2016-07-11)
Hippocampal synaptic plasticity and learning are regulated by metabotropic glutamate receptors (mGlu) and particularly by mGlu5. In the hippocampus, synaptic plasticity is tightly linked to neuronal network oscillations in theta (5-10 Hz) and gamma (∼30-100 Hz) frequency ranges, and specific changes in
A Ahnaou et al.
Psychopharmacology, 232(6), 1107-1122 (2014-10-18)
Evidence is emerging that positive and negative modulation of the metabotropic glutamate (mGluR5) receptors has the potential for treating cognitive deficits and neuroprotection associated with psychiatric and neurodegenerative diseases, respectively. Sleep and synchronisation of disparate neuronal networks are critically involved
Marta Marszalek-Grabska et al.
Behavioural brain research, 338, 9-16 (2017-10-17)
Repeated exposure to and withdrawal from ethanol induces deficits in spatial reversal learning. Data indicate that metabotropic glutamate 5 (mGlu5) receptors are implicated in synaptic plasticity and learning and memory. These receptors functionally interact with N-methyl-d-aspartate (NMDA) receptors, and activation
Jian Xu et al.
Learning & memory (Cold Spring Harbor, N.Y.), 20(8), 438-445 (2013-07-23)
Metabotropic glutamate receptor 5 (mGluR5) plays important roles in modulating neural activity and plasticity and has been associated with several neuropathological disorders. Previous work has shown that genetic ablation or pharmacological inhibition of mGluR5 disrupts fear extinction and spatial reversal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service