Skip to Content
Merck
All Photos(1)

Key Documents

SML2249

Sigma-Aldrich

LSN3154567

≥98% (HPLC)

Synonym(s):

2-Hydroxy-2-methyl-N-[1,2,3,4-tetrahydro-2-[2-(3-pyridinyloxy)acetyl]-6-isoquinolinyl]-1-propanesulfonamide, Nampt-IN-1; 2-Hydoxy-2-methyl-N-{2-[(pyridin-3-yloxy)acetyl]-1,2,3,4-tetrahydroisoquinolin-6-YL}propane-1-sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$564.00
25 MG
R$2,278.00

R$564.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$564.00
25 MG
R$2,278.00

About This Item

Empirical Formula (Hill Notation):
C20H25N3O5S
CAS Number:
Molecular Weight:
419.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$564.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc1cc2c(cc1)CN(CC2)C(=O)COc3cnccc3)CC(O)(C)C

InChI

1S/C20H25N3O5S/c1-20(2,25)14-29(26,27)22-17-6-5-16-12-23(9-7-15(16)10-17)19(24)13-28-18-4-3-8-21-11-18/h3-6,8,10-11,22,25H,7,9,12-14H2,1-2H3

InChI key

QHHSCLARESIWBH-UHFFFAOYSA-N

Biochem/physiol Actions

LSN3154567 is an orally available, potent and highly selective inhibitor of nicotinamide phosphoribosyltransferase (NAMPT). LSN3154567 exhibits broad spectrum and potent anticancer activities without causing retinopathy in the rat. LSN3154567co-administration with nicotinic acid (NA) produces potent anti-tumor activity in tumor xenograft models without retinal and hematological toxicities.
LSN3154567 might be useful in developing a novel cancer therapeutic.[1]
orally available, potent and highly selective inhibitor of nicotinamide phosphoribosyltransferase (NAMPT)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genshi Zhao et al.
Molecular cancer therapeutics, 16(12), 2677-2688 (2017-10-22)
NAMPT, an enzyme essential for NAD+ biosynthesis, has been extensively studied as an anticancer target for developing potential novel therapeutics. Several NAMPT inhibitors have been discovered, some of which have been subjected to clinical investigations. Yet, the on-target hematological and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service