Skip to Content
Merck
All Photos(1)

Documents

SML2208

Sigma-Aldrich

BRD6989

≥98% (HPLC)

Synonym(s):

2-Amino-3-cyano-6-methyl-4-(3-pyridinyl)-5,6,7,8-tetrahydroquinoline, 2-Amino-5,6,7,8-tetrahydro-6-methyl-4-(3-pyridinyl)-3-quinolinecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N4
CAS Number:
Molecular Weight:
264.33
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC(CCC(C)C2)=C2C(C3=CN=CC=C3)=C1C#N

Biochem/physiol Actions

BRD6989 is an inhibitor of cyclin-dependent kinase 8 (CDK8) that upregulates production of interleukin-10 (IL-10). BRD6989 was shown to bind cyclin C–CDK8 complex selectively with an IC50 value ∼200 nM and to inhibit the kinase activity of recombinant cyclin C–CDK8 or cyclin C–CDK19 complexes with IC50 values ∼500 nM and >30 μM, respectively. It enhanced the production interleukin-10 (IL-10) enhances IL-10 production in activated human and murine macrophages and dendritic cells with an EC50 value ∼1 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liv Johannessen et al.
Nature chemical biology, 13(10), 1102-1108 (2017-08-15)
Enhancing production of the anti-inflammatory cytokine interleukin-10 (IL-10) is a promising strategy to suppress pathogenic inflammation. To identify new mechanisms regulating IL-10 production, we conducted a phenotypic screen for small molecules that enhance IL-10 secretion from activated dendritic cells. Mechanism-of-action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service