Skip to Content
Merck
All Photos(1)

Key Documents

SML2207

Sigma-Aldrich

SR-4995

≥98% (HPLC)

Synonym(s):

CID 16016685, N-Butyl-N′-(10,11-dihydro-10-methyl-11-oxodibenzo[b,f][1,4]thiazepin-8-yl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$666.00
25 MG
R$2,696.00

R$666.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$666.00
25 MG
R$2,696.00

About This Item

Empirical Formula (Hill Notation):
C19H21N3O2S
CAS Number:
Molecular Weight:
355.45
UNSPSC Code:
12352200
NACRES:
NA.77

R$666.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CN1C(C(C=CC=C2)=C2SC3=C1C=C(NC(NCCCC)=O)C=C3)=O

InChI

1S/C19H21N3O2S/c1-3-4-11-20-19(24)21-13-9-10-17-15(12-13)22(2)18(23)14-7-5-6-8-16(14)25-17/h5-10,12H,3-4,11H2,1-2H3,(H2,20,21,24)

InChI key

UGYXUEPBYOKNOL-UHFFFAOYSA-N

Biochem/physiol Actions

SR-4995 is a potent and selective ligand of α-β-hydrolase domain containing 5 (ABHD5) that activates adipose triglyceride lipase (ATGL) by dissociating ABHD5 from its inhibitory regulator, perilipin-1 (PLIN1) and PLIN5. SR-4995 directly binds to ABHD5 and prevents ABHD5 to PLIN1. SR-4995 induces lipolysis in adipocytes and muscle, avoiding PKA-dependent signaling.
potent and selective ligand of ABHD5 that activates ATGL by dissociating ABHD5 from PLIN1 and PLIN5

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew A Sanders et al.
Cell metabolism, 22(5), 851-860 (2015-09-29)
Fat and muscle lipolysis involves functional interactions of adipose triglyceride lipase (ATGL), α-β hydrolase domain-containing protein 5 (ABHD5), and tissue-specific perilipins 1 and 5 (PLIN1 and PLIN5). ABHD5 potently activates ATGL, but this lipase-promoting activity is suppressed when ABHD5 is
Elizabeth A Rondini et al.
The Journal of pharmacology and experimental therapeutics, 363(3), 367-376 (2017-09-21)
Current knowledge regarding acute regulation of adipocyte lipolysis is largely based on receptor-mediated activation or inhibition of pathways that influence intracellular levels of cAMP, thereby affecting protein kinase A (PKA) activity. We recently identified synthetic ligands of α-β-hydrolase domain containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service