Skip to Content
Merck
All Photos(1)

Key Documents

SML2112

Sigma-Aldrich

GSK8815

≥98% (HPLC)

Synonym(s):

8-((3S,4S,5R)-3-((4,4-Difluorocyclohexyl)methoxy)-5-methoxypiperidin-4-ylamino)-3-methyl-5-(5-methylpyridin-3-yl)-1,7-naphthyridin-2(1H)-one, 8-[[(3S,4S,5R)-3-[(4,4-Difluorocyclohexyl)methoxy]-5-methoxy-4-piperidinyl]amino]-3-methyl-5-(5-methyl-3-pyridinyl)-1,7-naphthyridin-2(1H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$946.00
25 MG
R$3,785.00

R$946.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$946.00
25 MG
R$3,785.00

About This Item

Empirical Formula (Hill Notation):
C28H35F2N5O3
CAS Number:
Molecular Weight:
527.61
UNSPSC Code:
12352200
NACRES:
NA.77

R$946.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1)C(C)=CC2=C1C(N[C@H]3[C@H](OCC4CCC(F)(F)CC4)CNC[C@H]3OC)=NC=C2C5=CN=CC(C)=C5

Biochem/physiol Actions

GSK8815 is an aqueous soluble (326 μM) naphthyridone that serves as a negative control for the more active diastereomer GSK8814 in ATAD2/ATAD2B inhibition studies. Comparing to GSK8814, GSK8815 is ineffective in cell-based assays and exhibits greatly reduced affinity toward ATAD2 & ATAD2B bromodomain in cell-free binding assays (pKd for ATAD2 = 5.5/GSK8815 vs. 8.1/GSK8814 by ITC; pIC50 against ATAD2 BD/ATAD2B BD = 5.5/5.5 with GSK8815 vs. 7.3/7.7 with GSK8814 in competitive ligand binding assays by TR-FRET).
Less potent control compound for GSK8814

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Bamborough et al.
Angewandte Chemie (International ed. in English), 55(38), 11382-11386 (2016-08-18)
ATAD2 is a cancer-associated protein whose bromodomain has been described as among the least druggable of that target class. Starting from a potent lead, permeability and selectivity were improved through a dual approach: 1) using CF2 as a sulfone bio-isostere

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service