Skip to Content
Merck
All Photos(1)

Key Documents

SML2032

Sigma-Aldrich

Win6mer trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

Ac-ARTEVY-NH2 trifluouroaceate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$1,110.00

R$1,110.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
R$1,110.00

About This Item

Empirical Formula (Hill Notation):
C34H54N10O11 · xC2HF3O2
CAS Number:
Molecular Weight:
778.85 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

R$1,110.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

film (Clear)

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@H](NC(C)=O)C(N[C@H](C(N[C@@](C(N[C@H](C(N[C@H](C(N[C@H](C(N)=O)CC1=CC=C(C=C1)O)=O)C(C)C)=O)CCC(O)=O)=O)([H])[C@H](O)C)=O)CCCNC(N)=N)=O

InChI

1S/C34H54N10O11/c1-16(2)26(32(54)42-24(28(35)50)15-20-8-10-21(47)11-9-20)43-31(53)23(12-13-25(48)49)41-33(55)27(18(4)45)44-30(52)22(7-6-14-38-34(36)37)40-29(51)17(3)39-19(5)46/h8-11,16-18,22-24,26-27,45,47H,6-7,12-15H2,1-5H3,(H2,35,50)(H,39,46)(H,40,51)(H

InChI key

DYNVFAUYAHJESK-NGPBCSSISA-N

Biochem/physiol Actions

Win6mer is a potent and specific inhibitor of MLL1 and SETd1A histone H3 lysine 4 (H3K4) methyltransferase complexes that does not inhibit MLL2-4 or SETd1B complexes. Win6mer potently binds to WDR5 in a 310-helical conformation. It does not inhibit isolated SET domain activity.
potent and specific inhibitor of MLL1 and SETd1A histone H3 lysine 4 (H3K4) methyltransferase complexes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nilda L Alicea-Velázquez et al.
The Journal of biological chemistry, 291(43), 22357-22372 (2016-08-27)
MLL1 belongs to the SET1 family of histone H3 lysine 4 (H3K4) methyltransferases, composed of MLL1-4 and SETd1A/B. MLL1 translocations are present in acute leukemias, and mutations in several family members are associated with cancer and developmental disorders. MLL1 associates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service