Skip to Content
Merck
All Photos(1)

Key Documents

SML1915

Sigma-Aldrich

T-26c

≥98% (HPLC)

Synonym(s):

4-[[[1,4-Dihydro-2-[[[(3-methoxyphenyl)methyl]amino]carbonyl]-4-oxothieno[2,3-d]pyrimidin-5-yl]methoxy]methyl]-benzoic acid; 4-[[[[2-[[[[3-(Methyloxy)phenyl]methyl]amino]carbonyl]-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidin-5-yl]methyl]oxy]methyl]benzoic acid, 4-[[[[2-[[[[3-(Methyloxy)phenyl]methyl]amino]carbonyl]-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidin-5-yl]methyl]oxy]methyl]benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$693.00
25 MG
R$2,227.00

R$693.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$693.00
25 MG
R$2,227.00

About This Item

Empirical Formula (Hill Notation):
C24H21N3O6S
CAS Number:
Molecular Weight:
479.51
UNSPSC Code:
41106609
NACRES:
NA.77

R$693.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C(C=C1)=CC=C1COCC2=CSC3=C2C(NC(C(NCC4=CC=CC(OC)=C4)=O)=N3)=O)=O

Biochem/physiol Actions

T-26c is a highly potent, selective and cell-permeable matrix metalloproteinases 13 (MMP-13) inhibitor.
T-26c is a highly potent, selective and cell-permeable matrix metalloproteinases 13 (MMP-13) inhibitor. T-26c inhibits the breakdown of collagen (87.4% inhibition at 0.1 μM) in IL-1b and oncostatin M stimulated cartilage. For full characterization details, please visit the T-26c probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for protein targets, visit sigma.com/sgc

Features and Benefits

T-26c is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Nara et al.
Bioorganic & medicinal chemistry, 22(19), 5487-5505 (2014-09-07)
On the basis of X-ray co-crystal structures of matrix metalloproteinase-13 (MMP-13) in complex with its inhibitors, our structure-based drug design (SBDD) strategy was directed to achieving high affinity through optimal protein-ligand interaction with the unique S1″ hydrophobic specificity pocket. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service