Skip to Content
Merck
All Photos(1)

Key Documents

SML1842

Sigma-Aldrich

Ibuprofen Arginate

≥98% (HPLC)

Synonym(s):

α-Methyl-4-(2-methylpropyl)benzeneacetate, L-arginine (1:1), (S)-1-(4-Amino-4-carboxybutyl)guanidinium 2-(4-isobutylphenyl)propanoate, Ibu-L-Arg, Ibuprofen L-arginine, Ibuprofen arginine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$2,090.00

R$2,090.00


Check Cart for Availability


Select a Size

Change View
25 MG
R$2,090.00

About This Item

Empirical Formula (Hill Notation):
C19H32N4O4
CAS Number:
Molecular Weight:
380.48
UNSPSC Code:
51111800
NACRES:
NA.77

R$2,090.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(CC1=CC=C(C(C([O-])=O)C)C=C1)C.O=C(O)[C@@H](N)CCCNC([NH3+])=N

InChI

1S/C13H18O2.C6H14N4O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;7-4(5(11)12)2-1-3-10-6(8)9/h4-7,9-10H,8H2,1-3H3,(H,14,15);4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t;4-/m.0/s1

InChI key

GCCOJNYCFNSJII-VWMHFEHESA-N

Biochem/physiol Actions

Ibuprofen arginate is the more soluble arginate salt form of the nonsteroidal anti-inflammatory drug (NSAID) ibuprofen.
Ibuprofen arginate is the more soluble arginate salt form of the nonsteroidal anti-inflammatory drug (NSAID) ibuprofen. Ibuprofen arginate retains the inhibitory potency against COX-2 as ibuprofen and its sodium salt, however only ibuprofen arginate is able to augment cellular NO pathway through codelivery of the NOS substrate arginine. Ibuprofen arginate, but not the sodium salt form, is shown to reverse the inhibitory effects of the cardiotoxic hormone asymmetric dimethylarginine (ADMA) and NG-nitro-L-argininemethyl ester (L-NAME) against nitric oxide synthase (eNOS & iNOS) in cultures in vitro (Murine RAW264.7 macrophages and isolated mouse & rat aorta) and effectively prevent L-NAME-induced arterial blood pressure increase in anesthetized rats in vivo (0.5 mg/kg L-NAME; 114.8 μmol/kg ibuprofen arginate, equivalent to 20 mg/kg L-Arg via femoral artery cannulation).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lobna A Saleh et al.
Pharmacology, biochemistry, and behavior, 125, 29-39 (2014-08-08)
Prostaglandins (PGs) and nitric oxide (NO) may be involved in the pathophysiology of depression. Since NSAIDs decrease PGs and NO production, they may have an antidepressant effect. The aim of the present work was to explore a possible antidepressant action
Ghada M El-Lithy et al.
Translational research : the journal of laboratory and clinical medicine, 177, 85-97 (2016-07-10)
Diabetic neuropathy (DN) is a common complication of diabetes mellitus that is hardly reversible at the late stages. Since treatment of neuropathic pain is predominantly symptomatic, a prophylactic measure would be useful. Both ibuprofen and L-arginine exert antiallodynic effects on
Pharmacological assessment of ibuprofen arginate on platelet aggregation and colon cancer cell killing.
Ahmetaj-Shala, B., et al.
Biochemical and Biophysical Research Communications, 18484(4), 762-766 (2017)
Nicholas S Kirkby et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(12), 4172-4179 (2016-09-08)
Nonsteroidal antiinflammatory drugs, including ibuprofen, are among the most commonly used medications and produce their antiinflammatory effects by blocking cyclooxygenase (COX)-2. Their use is associated with increased risk of heart attacks caused by blocking COX-2 in the vasculature and/or kidney

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service